Merck
Identification of a novel mechanism for the removal of glucose residues from high mannose-type oligosaccharides.
The Journal of biological chemistry (1992-10-25)
K Suh, C A Gabel, J E Bergmann
ABSTRAKT
The role of glucosylated oligosaccharides in the biogenesis of the glycoprotein (G protein) of vesicular stomatitis virus was studied in PhaR2.7, a mouse lymphoma cell line deficient in glucosidase II activity. As expected, the great majority of cell-associated G protein remained glucosylated in PhaR2.7, and the G protein was rapidly deglucosylated in BW5147, the parental cell line. Despite these differences in glucosylation, the rates of G protein trimerization and transport to the cell surface were as rapid and efficient in the PhaR2.7 mutant as in BW5147. Surprisingly, greater than 73% of the oligosaccharides on G proteins recovered from released virions were complex-type units. The efficient processing of the G protein oligosaccharides coincided with the efficient removal of glucose residues from its oligosaccharides. After treatment with deoxynojirimycin, an inhibitor of endoplasmic reticulum (ER) glucosidases I and II, the total percentage of G protein-associated high mannose-type oligosaccharides increased more in the parental cells than in the mutant cells. Furthermore, when the G protein was retained in the ER of PhaR2.7 cells by depletion of the cellular ATP pools with carbonyl cyanide m-chlorophenylhydrazone, its oligosaccharides remained glucosylated. Under identical conditions, BW5147 cells removed the glucose residues from > 90% of the retained G protein's oligosaccharides. Thus, PhaR2.7 cells efficiently remove glucose residues from high mannose-type oligosaccharides of selected proteins using a deoxynojirimycin-insensitive enzyme located in a post-ER compartment. The existence of a second mechanism for the deglucosylation of N-linked oligosaccharides provides evidence for the important role of glucose removal in glycoprotein maturation.