Merck
Urocortin 2 lowers blood pressure and reduces plasma catecholamine levels in mice with hyperadrenergic activity.
Endocrinology (2010-07-30)
Yusu Gu, Kuixing Zhang, Nilima Biswas, Ryan S Friese, Dennis H Lin, Sushil K Mahata, Masahiko Hoshijima, Daniel T O'Connor, Kirk L Peterson, Bhawanjit K Brar
ABSTRAKT
Exaggerated adrenergic activity is associated with human hypertension. The peptide urocortin 2 (Ucn 2) inhibits catecholamine synthesis and secretion from adrenal chromaffin cells in vitro and administration to mammals lowers blood pressure (BP). The chromogranin A-null mouse (Chga-/-) manifests systemic hypertension because of excessive catecholamine secretion from the adrenal and decreased catecholamine storage. In the present study, we investigated whether systemic administration of Ucn 2 could reduce BP and adrenal and plasma levels of catecholamines in vivo. Ucn 2 peptide was administered to freely moving, conscious Chga-/- and wild-type control mice. Telemetry and HPLC measured changes in BP and catecholamine levels, respectively. In both groups of mice, Ucn 2 dose-dependently decreased BP, and this effect was mediated by corticotropin factor-receptor type 2. However, in Chga-/- mice, the maximal percentage decrease of systolic BP from basal systolic BP was 37% compared with only a 23% reduction in wild-type mice (P=0.04). In Chga-/- mice only, Ucn 2 decreased adrenal and plasma levels of catecholamines as well as adrenal levels of tyrosine hydroxylase protein and phosphorylation. In vitro mechanistic studies demonstrated that Ucn 2 reduces both catecholamine secretion and tyrosine hydroxylase promoter activity, suggesting that the exaggerated action of Ucn 2 to reduce BP in the Chga-/- mouse is mediated through inhibition of both catecholamine synthesis and secretion. The data suggest that Ucn 2 may be therapeutically useful in regulating the exaggerated sympathoadrenal function of hyperadrenergic hypertension.