Merck
On the role of helper lipids in lipid nanoparticle formulations of siRNA.
Nanoscale (2019-11-13)
Jayesh A Kulkarni, Dominik Witzigmann, Jerry Leung, Yuen Yi C Tam, Pieter R Cullis
ABSTRAKT
Onpattro, the first RNAi-based therapeutic to receive FDA approval, is enabled by a lipid nanoparticle (LNP) system that facilitates siRNA delivery into the cytoplasm of target cells (hepatocytes) following intravenous (i.v.) administration. These LNP-siRNA systems consist of four lipid components (ionizable cationic lipid, distearolyphosphatidycholine or DSPC, cholesterol, and PEG-lipid) and siRNA. The ionizable cationic lipid has been optimised for RNA encapsulation and intracellular delivery, and the PEG-lipids have been engineered to regulate LNP size and transfection potency. The roles of the other "helper" lipids, DSPC and cholesterol, remain less clear. Here we show that in empty LNP systems that do not contain siRNA, DSPC-cholesterol resides in outer layers, whereas in loaded systems a portion of the DSPC-cholesterol is internalised together with siRNA. It is concluded that the presence of internalised helper lipid is vital to the stable encapsulation of siRNA in the LNP and thus to LNP-siRNA function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE (Ammonium Salt), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Triolein, European Pharmacopoeia (EP) Reference Standard
Avanti
Egg SM, Avanti Research - A Croda Brand
Avanti
Egg SM, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand