Merck
Portal Angiogenesis in Chronic Liver Disease Patients Correlates with Portal Pressure and Collateral Formation.
Digestive diseases (Basel, Switzerland) (2019-05-09)
Carolina A Serrano, Simon C Ling, Sofia Verdaguer, Miguel León, Nicolás Jarufe, Juan Francisco Guerra, Juan Carlos Pattillo, Carlos Benítez, Andrea Villagrán, Javiera Torres, Mario Concha, Luis Villarroel, Paulina Dellepiane, Pilar Domínguez, Jorge Martínez, Juan Cristóbal Gana
ABSTRAKT
One hallmark of chronic liver disease in patients with portal hypertension is the formation of portal-systemic collaterals in which angiogenesis has a fundamental role. We studied patients with chronic liver disease undergoing liver transplantation to correlate levels of circulating angiogenic factors in portal and peripheral circulation with portal pressure and portal-systemic collaterals. Sixteen patients who underwent liver transplantation were enrolled. During transplant surgery, we determined portal venous pressure and portal-systemic collateral formation. We determined angiogenics mediator levels in systemic and portal plasma. Peripheral plasma from healthy donors was measured as controls. Vascular endothelial growth factor (VEGF)-R1 and 2, Ang-1 and 2, Tie2, FGF- 1 and 2, CD163, PDGFR-β, PDGFsRα, PDGF-AB and BB, CD163, TGF-β VASH-1 levels were significantly different in the controls in comparison to cases. Significantly decreased portal venous levels of Ang-1, FGF-1, PDGF-AB/BB, and CC were observed in patients with higher portal pressure. Peripheral VEGF, Ang-1, pPDGF-AB, BB, and CC were significantly decreased in patients with more severe collateral formation. While peripheral VEGF-R1 was higher in patients with severe collateral formation. For portal circulation, VEGF, Ang-1, -pPDGF-AB, BB, and CC were significantly decreased in patients with more severe collateral formation Conclusions: Angiogenesis factors correlated with portal pressure and collateral formation and different patterns of circulating angiogenesis mediators were found in peripheral and portal blood of patients with chronic liver disease. These results support the importance of angiogenic pathways in cirrhosis and portal hypertension and highlight areas for further study to identify clinically useful noninvasive markers of portal pressure and collateral formation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder