Merck
Protective Efficacy of Nucleic Acid Vaccines Against Transmission of Zika Virus During Pregnancy in Mice.
The Journal of infectious diseases (2019-07-02)
Brett W Jagger, Kimberly A Dowd, Rita E Chen, Pritesh Desai, Bryant Foreman, Katherine E Burgomaster, Sunny Himansu, Wing-Pui Kong, Barney S Graham, Theodore C Pierson, Michael S Diamond
ABSTRAKT
Zika virus (ZIKV) caused an epidemic of congenital malformations in 2015-2016. Although many vaccine candidates have been generated, few have demonstrated efficacy against congenital ZIKV infection. Here, we evaluated lipid-encapsulated messenger RNA (mRNA) vaccines and a DNA plasmid vaccine encoding the prM-E genes of ZIKV in mouse models of congenital infection. Although the DNA vaccine provided comparable efficacy against vertical transmission of ZIKV, the mRNA vaccines, including one that minimizes antibody-dependent enhancement of infection, elicited higher levels of antigen-specific long-lived plasma cells and memory B cells. Despite the induction of robust neutralizing antibody titers by all vaccines, breakthrough seeding of the placenta and fetal head was observed in a small subset of type I interferon signaling-deficient immunocompromised dams. In comparison, evaluation of one of the mRNA vaccines in a human STAT2-knockin transgenic immunocompetent mouse showed complete protection against congenital ZIKV transmission. These data will inform ongoing human ZIKV vaccine development efforts and enhance our understanding of the correlates of vaccine-induced protection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~2,500 IU