Merck
Lithium Titanate Cuboid Arrays Grown on Carbon Fiber Cloth for High-Rate Flexible Lithium-Ion Batteries.
Small (Weinheim an der Bergstrasse, Germany) (2019-08-29)
Chao Wang, Xianfen Wang, Chunfu Lin, Xiu Song Zhao
ABSTRAKT
High-rate performance flexible lithium-ion batteries are desirable for the realization of wearable electronics. The flexibility of the electrode in the battery is a key requirement for this technology. In the present work, spinel lithium titanate (Li4 Ti5 O12 , LTO) cuboid arrays are grown on flexible carbon fiber cloth (CFC) to fabricate a binder-free composite electrode (LTO@CFC) for flexible lithium-ion batteries. Experimental results show that the LTO@CFC electrode exhibits a remarkably high-rate performance with a capacity of 105.8 mAh g-1 at 50C and an excellent electrochemical stability against cycling (only 2.2% capacity loss after 1000 cycles at 10C). A flexible full cell fabricated with the LTO@CFC as the anode and LiNi0.5 Mn1.5 O4 coated on Al foil as the cathode displays a reversible capacity of 109.1 mAh g-1 at 10C, an excellent stability against cycling and a great mechanical stability against bending. The observed high-rate performance of the LTO@CFC electrode is due to its unique corn-like architecture with LTO cuboid arrays (corn kernels) grown on CFC (corn cob). This work presents a new approach to preparing LTO-based composite electrodes with an architecture favorable for ion and electron transport for flexible energy storage devices.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lithium titanate, −80 mesh
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)