Merck
  • Development of a lipoplex-type mRNA carrier composed of an ionizable lipid with a vitamin E scaffold and the KALA peptide for use as an ex vivo dendritic cell-based cancer vaccine.
Development of a lipoplex-type mRNA carrier composed of an ionizable lipid with a vitamin E scaffold and the KALA peptide for use as an ex vivo dendritic cell-based cancer vaccine.
Journal of controlled release : official journal of the Controlled Release Society (2019-08-07)
Naho Tateshita, Naoya Miura, Hiroki Tanaka, Takeshi Masuda, Sumio Ohtsuki, Kota Tange, Yuta Nakai, Hiroki Yoshioka, Hidetaka Akita
ABSTRAKT
A dendritic cells (DCs)-based vaccine (DC-vaccine) system is an attractive technology for eliciting antigen-specific immune responses that can protect subjects from infectious diseases and for curing various types of cancers. For the insertion of a foreign antigen to DCs, the transfection of an antigen-coding mRNA to the cells is a promising approach. In order to introduce an antigen, a carrier for mRNA transfection is required, since the mRNA molecule per se is unstable in serum-containing medium. We previously reported on an ionizable lipid-like material with vitamin E-scaffolds (ssPalmE) as a material for a lipid nanoparticle (LNP)-based carrier for nucleic acids. In the present study, we report on the development of a lipoplex-type mRNA carrier for use as a DC-vaccine by using a combination of an ssPalmE-LNP and an α-helical cationic peptide "KALA" (ssPalmE-KALA). The transfection of mRNAs complexed with the ssPalmE-KALA achieved a significantly higher protein expression and the production of proinflammatory cytokines from murine bone marrow derived DCs (BMDCs) in comparison with a lipoplex that was prepared with an ssPalm with fatty acid-scaffolds (myristic acid; ssPalmM-KALA). A cellular uptake process and a pH-responsive membrane-destabilization activity cannot explain the preferred protein expression and immune-stimulation caused by the ssPalmE-KALA. Proteomic analyses suggest that transfection with the ssPalmM-KALA stimulates a down-regulatory pathway of translation, while the transfection with the ssPalmE-KALA does not stimulate it. In the vaccination with the BMDCs that were preliminarily transfected with an ovalbumin (OVA)-encoding mRNA elicited the induction OVA specific cytotoxic T-lymphocyte activity in vivo. In parallel, the vaccination induced significant prophylactic anti-tumor effects against a model tumor that stably expressed the OVA protein. Based on the above findings, the ssPalmE-KALA appears to be a potent ex vivo DCs-based RNA vaccine platform.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder