Merck
Abiotic stress modulates root patterning via ABA-regulated microRNA expression in the endodermis initials.
Development (Cambridge, England) (2019-08-11)
Daria Bloch, Malikarjuna Rao Puli, Assaf Mosquna, Shaul Yalovsky
ABSTRAKT
Patterning of the root xylem into protoxylem (PX) and metaxylem is regulated by auxin-cytokinin signaling and microRNA miR165a/166b-mediated suppression of genes encoding Class III HOMEODOMAIN LEU-ZIPPER (HD-ZIPIII) proteins. We found that, in Arabidopsis, osmotic stress via core abscisic acid (ABA) signaling in meristematic endodermal cells induces differentiation of PX in radial and longitudinal axes in association with increased VND7 expression. Similarly, in tomato, ABA enhanced PX differentiation longitudinally and radially, indicating an evolutionarily conserved mechanism. ABA increased expression of miR165a/166b and reduced expression of the miR165a/166b repressor ZWILLE (ZLL) (also known as ARGONAUTE10), resulting in reduced levels of all five HD-ZIPIII RNAs. ABA treatments failed to induce additional PX files in a miR165a/166b-resistant PHB mutant, phb1-d, and in scr and shr mutants, in which miR165a/166b expression is strongly reduced. Thus, ABA regulates xylem patterning and maturation via miR165a/166b-regulated expression of HD-ZIPIII mRNAs and associated VND7 levels. In lateral root initials, ABA induced an increase in miR165a levels in endodermal precursors and inhibited their reduction in the future quiescent center specifically at pre-emergence stage. Hence, ABA-induced inhibition of lateral root is associated with reduced HD-ZIPIII levels.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Supelco
Mandipropamid, PESTANAL®, analytical standard
Sigma-Aldrich
Calcofluor White Stain, suitable for microbiology
Sigma-Aldrich
Poly(ethylene glycol), Molecular Biology, average mol wt 8,000
Sigma-Aldrich
Propidium iodide solution