Merck
Ubiquitin-specific peptidase 3 induces TPA-mediated leukemia cell differentiation via regulating H2AK119ub.
Animal cells and systems (2019-11-09)
Yun-Cheol Chae, Hyeonsoo Jung, Ji-Young Kim, Dong Ho Lee, Sang-Beom Seo
ABSTRAKT
Acute myeloid leukemia (AML) is the most common type of leukemia in adults. Owing to the chemotherapy associated side effects and toxicity, it is necessary to find a new mechanism, which can identify new potential therapeutic targets at the molecular level. Here, we identified new target genes that are induced during the TPA-induced HL-60 cell differentiation by ChIP-seq and microarray data analysis. Using q-PCR and ChIP assay, we confirmed that the target genes including USP3, USP35, TCF4, and SGK1 are upregulated during TPA-mediated HL-60 cell differentiation. Levels of USP3, one of the deubiquitinating enzymes (DUBs), increased by TPA treatment, resulting in the reduction of H2AK119ub levels. In addition, we revealed that depletion of USP3 inhibits TPA-mediated leukemia cell differentiation q-PCR and FACS analysis. Taken together, our data indicate that USP3 promotes TPA-mediated leukemia cell differentiation via regulating H2AK119ub levels.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human USP3
Sigma-Aldrich
Przeciwciało anty-histonowe H3, klon 6.6.2, clone 6.6.2, Upstate®, from mouse
Sigma-Aldrich
Przeciwciało anty-ubikwitylo-histonowe H2A, klon E6C5, clone E6C5, Upstate®, from mouse