Merck
  • Desmoglein 3 gene mediates epidermal growth factor/epidermal growth factor receptor signaling pathway involved in inflammatory response and immune function of anaphylactic rhinitis.
Desmoglein 3 gene mediates epidermal growth factor/epidermal growth factor receptor signaling pathway involved in inflammatory response and immune function of anaphylactic rhinitis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2019-08-06)
Han Ri, Zheng Peiyan, Wang Jianqi, Zhao Yunteng, Li Gang, Sun Baoqing
ABSTRAKT
To investigate the effects of desmoglein 3 (DSG3) gene mediating epidermal growth factor/epidermal growth factor receptor (EGF/EGFR) signaling pathway on inflammatory response and immune function of anaphylactic rhinitis (AR). Ten of the seventy male BALB/c mice were randomly selected as the normal control group, and the remaining 60 were used to construct the AR mice model. AR model mice were divided into 6 groups: model group (instilled with 5 μL saline), empty vector group (instilled with 5 μL of liposome and empty vector mixture), siRNA-DSG3 group (instilled with 5 μL of liposome and siRNA-DSG3 carrier mixture), AG1478 group (instilled with 5 μL of EGF/EGFR inhibitor AG1478), siRNA-DSG3+AG1478 group (instilled with 5 μL of liposome and siRNA-DSG3 carrier and EGF/EGFR inhibitor AG1478 mixture) and oe-DSG3 group, 10 in each group. After taking serum, each group of mice was sacrificed to get nasal mucosa tissues. HE staining was used to observe the pathological changes of nasal mucosa tissues in each group. The expression levels of DSG3, EGF and EGFR in nasal mucosa tissues of mice in each group were detected by qRT-PCR and western blot methods respectively. TUNEL staining was used to observe the apoptosis of nasal mucosa cells in mice. The expression of IgE, INF-γ, TNF-α, IL-2, IL-4 and IL-6 in serum of mice was determined by ELISA method. The immune adhesion function of red blood cells was detected by complement sensitization yeast hemagglutination method. All the mice with AR showed different degrees of nasal mucosa injury and inflammatory cell infiltration, and silencing DSG3 or inhibiting the activity of EGF signaling pathway could alleviate the nasal mucosa injury. Compared with control group, the INF-γ and IL-2 levels of serum in AR model mice were significantly decreased; IgE, TNF-α, IL-4 and IL-6 levels were significantly increased (all P < 0.05); the mRNA expression levels and protein levels of DSG3, EGF and EGFR were significantly increased (all P < 0.05); C3b receptor rosette rate and Ic rosette rate were significantly decreased (all P < 0.05). Detected by ELISA method, the expression levels of IgE, TNF-α, IL-4 and IL-6 were increased, while the expression levels of INF-γ and IL-2 were decreased after DSG3 silencing or using AG1478. Detected by qRT-PCR and western blot methods, the expression of DSG3, EGF and EGFR did decrease after DSG3 silencing. There was no significant difference in the EGF and EGFR expression between DSG3 silencing and using AG1478, and the expression decreased even more under the double effect. The mRNA and protein expression levels of DSG3, EGF and EGFR in the nasal mucosa tissues of mice with overexpression of DSG3 plasmid were significantly higher than those of normal mice (all P < 0.05). Silencing DSG3 gene can inhibit the activation of EGF signaling pathway, alleviate the inflammation of AR nasal mucosa, and enhance red blood cells immune adherence function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human DSG3