Merck
An inkblot for beliefs: The Truth Misattribution Procedure.
PloS one (2019-06-25)
Jamie Cummins, Jan De Houwer
ABSTRAKT
An increasing body of evidence shows the importance of accommodating relational information within implicit measures of psychological constructs. Whereas relational variants of the Implicit Association Test (IAT) have been proposed in the past, we put forward the Truth Misattribution Procedure (TMP) as a relational variant of the Affect Misattribution Procedure (AMP) that aims to capture implicit beliefs. Across three experiments, we demonstrate that TMP effects are sensitive to the relational information contained within sentence primes, both in the context of causal stimulus relations of a known truth value (e.g., "smoking causes cancer" vs. "smoking prevents cancer"), as well as in the domain of gender stereotypes (e.g., "men are arrogant" vs. "men should be arrogant"). The potential benefits of the TMP are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thymidine 5′-monophosphate disodium salt hydrate, ≥99%