Merck
Isolation and culture of epithelial cells from stored buffalo semen and their use for the production of cloned embryos.
Reproduction, fertility, and development (2019-06-13)
Monika Saini, Naresh L Selokar, Rasika Rajendran, Dharmendra Kumar, Pradeep Kumar, Prem Singh Yadav
ABSTRAKT
The aim of the present study was to isolate somatic cells from semen, a non-invasive source of donor somatic cells, for somatic cell nuclear transfer (SCNT) experiments. The study had two parts: (1) isolation and culture of somatic cells from semen, which was stored at 4°C; and (2) investigating the SCNT competence of semen-derived somatic cells. We successfully cultured somatic cells from freshly ejaculated semen, which was stored for different times (0, 4, 12, 24, 72 and 144h after semen collection) at 4°C, using a Percoll gradient method. Up to 24h storage, 100% cell attachment rates were observed; cell attachment rates of 66% were observed for the 72 and 144h storage groups. The attached cells observed in all groups examined were proliferated (100%). Cultured cells exhibited epithelial cell morphology and culture characteristics, which was further confirmed by positive expression of cytokeratin 18, an epithelial cell-type marker. We compared the SCNT competence of semen-derived epithelial cells and skin-derived fibroblasts. The cleavage rate, blastocyst production rate, total number of cells in blastocysts and the apoptotic index of blastocysts were similar for embryos produced from semen-derived epithelial cells and skin-derived fibroblasts, indicating that semen-derived epithelial cells can serve as donors for SCNT experiments. In conclusion, we demonstrate a method to culture epithelial cells from stored semen, which can be used to produce cloned embryos of breeding bulls, including remote bulls.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Medium 199, HEPES Modification, with Earle′s salts, 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture