Merck
Immobilized lignin peroxidase on Fe3O4@SiO2@polydopamine nanoparticles for degradation of organic pollutants.
International journal of biological macromolecules (2019-07-22)
Jin Guo, Xiuchu Liu, Xiaomei Zhang, Juan Wu, Chao Chai, Dong Ma, Qinghua Chen, Dan Xiang, Wei Ge
ABSTRAKT
Lignin peroxidase (LiP) was obtained from Pichia methanolica through heterologous expression. LiP was extracted, purified, and immobilized on Fe3O4@SiO2@polydopamine (PDA) nanoparticles to acquire immobilized LiP. The optimal preparation conditions for immobilized LiP were investigated. Results showed that the immobilization efficiency of immobilized LiP reached 56.37% when the enzyme amount, PDA concentration, and immobilization time were 12 mg, 1.6 mg/mL, and 12 h, respectively. Compared with free LiP, the immobilized LiP showed good thermal stability and storage stability and improved pH tolerance. It also retained more than 30% of its initial activity after 8 cycles, demonstrating its improved reusability. The immobilized LiP demonstrated efficacy of reaction of 100%, 100%, 100%, 100%, 79%, 73%, and 65% for tetracycline, dibutyl phthalate, 5-chlorophenol, phenol, phenanthrene, fluoranthene, and benzo(a)pyrene, respectively, while the inactivated immobilized LiP only adsorbed <25% of phenanthrene and fluoranthene. The dissipation of organic pollutants was a combination of degradation and adsorption, with the former playing a more important role.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyrene, 98%