Merck
Validating an ion mobility spectrometry-quadrupole time of flight mass spectrometry method for high-throughput pesticide screening.
The Analyst (2019-07-11)
Xiu-Ping Chen, Fang Zhang, Yin-Long Guo
ABSTRAKT
The utility of adding ion mobility (IM) to quadrupole time of flight mass spectrometry (IM-QTOF MS) for highly effective analysis of multiple pesticides in complex matrices was evaluated. Based on an in-house IM-MS database, the identification was performed through the match of the protonated ion ([M + H]+) and the CCS value. Moreover, the structural confirmation was achieved by using the accurate masses of [M + H]+ with its fragment ions, and the reference CCS value. The method did not require chromatographic separation and the analysis time of each measurement cycle is 1.6 min. The "cleaned" IM-MS spectra afforded by the drift time filtration improved the reliability of structural confirmation. As a result, the limit of detection (LOD) of 92% of test pesticides under the APCI mode and 58% of test pesticides under the ESI mode spiked in scallion was not more than 20 ng mL-1. In the analysis of practical samples, the identification of pyrimethanil was confirmed in celery, and benalaxyl and tebuconazole were identified as false positives in scallion. The time-saving, extended-scope and high-throughput method described in this work is capable of determining multiple pesticide residues in complex matrices with high sensitivity for monitoring applications.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Pretilachlor, PESTANAL®, analytical standard
Supelco
Cyprodinil, PESTANAL®, analytical standard