Merck
  • Photochemical or electrochemical bond breaking - exploring the chemistry of (μ2-alkyne)Co2(CO)6 complexes using time-resolved infrared spectroscopy, spectro-electrochemical and density functional methods.
Photochemical or electrochemical bond breaking - exploring the chemistry of (μ2-alkyne)Co2(CO)6 complexes using time-resolved infrared spectroscopy, spectro-electrochemical and density functional methods.
Dalton transactions (Cambridge, England : 2003) (2019-09-10)
Jennifer C Manton, Florian J R Cerpentier, Emma C Harvey, Ian P Clark, Gregory M Greetham, Conor Long, Mary T Pryce
ABSTRAKT
The photochemistry of (μ2-CRCR')Co2(CO)6 complexes (R = pyrenyl, R' = H; R = pyrenyl, R' = ferrocenyl; R = ferrocenyl, R = H) was investigated by ps-time-resolved infrared spectroscopy at room temperature in dichloromethane solution. The main focus of these studies was to determine the primary photoprocess relevant to the light assisted Pauson-Khand reaction. These studies were supported by spectro-electrochemical investigations and density functional calculations which suggest that the primary process to initiate the Pauson-Khand reaction involves a homolytic cleavage of the Co-Co bond forming a high-spin diradical species and not CO-loss as previously thought.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Bromopyrene, 96%
Sigma-Aldrich
Ethynyltrimethylsilane, 98%