Merck
Determination of Selected Opiates in Hair Samples Using Microextraction by Packed Sorbent: A New Approach for Sample Clean-up.
Journal of analytical toxicology (2019-07-23)
Tiago Rosado, Mário Barroso, Duarte Nuno Vieira, Eugenia Gallardo
ABSTRAKT
In this article the development and validation of an analytical method using microextraction by packed sorbent (MEPS) to determine tramadol (TRM), codeine (COD), morphine (MOR), 6-acetylcodeine (6-AC), 6-monoacetylmorphine (6-MAM) and fentanyl (FNT) in hair samples by gas chromatography coupled to tandem mass spectrometry (GC-MS-MS) is presented. The MEPS used a mixed mode sorbent, and the steps for sample cleanup were conditioning (three cycles of 250 μL of methanol and three cycles of 250 μL formic acid 2%); sample load (15 cycles of 150 μL); wash (150 μL of 3.36% formic acid); and elution (eight cycles of 100 μL of ammonium hydroxide 2.36% (in methanol)). Linearity was obtained from the lower limit of quantitation (LLOQ) up to 5 ng/mg, with all target compounds revealing determination coefficients >0.99. The LLOQs achieved were 0.01 ng/mg for TRM, COD and 6-AC, and 0.025 ng/mg for MOR, 6-MAM and FNT. The recoveries ranged from 74 to 90% (TRM), 51 to 59% (COD), 22 to 36% (MOR), 69 to 99% (6-AC), 53 to 61% (6-MAM) and 75 to 86% (FNT). Precision and accuracy revealed coefficients of variation typically below 15% and relative errors within a ±15% interval, respectively. This new approach has proven to be an excellent alternative to classic procedures, reducing the volumes of organic solvents required.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propylene sulfite, 99%