Merck
  • Fabrication, Mechanisms, and Properties of High-Performance Flexible Transparent Conductive Gas-Barrier Films Based on Ag Nanowires and Atomic Layer Deposition.
Fabrication, Mechanisms, and Properties of High-Performance Flexible Transparent Conductive Gas-Barrier Films Based on Ag Nanowires and Atomic Layer Deposition.
ACS applied materials & interfaces (2019-08-30)
Dung-Yue Su, Che-Chen Hsu, Wen-Hsuan Lai, Feng-Yu Tsai
ABSTRAKT
Thin films of Ag nanowires (NWs) offer many advantages as transparent electrodes for flexible electronics, but their applications are hindered by issues including poor stability/durability of Ag NWs, high processing temperatures, heterogeneity of surfaces, and lack of gas-barrier function. This study reports novel mechanisms through which a conductive Hf:ZnO (HZO) film by atomic layer deposition (ALD) can be integrated with a sprayed Ag NWs film to address the issues of Ag NWs. First, the ALD surface reactions can induce fusing of the Ag NWs into a connected network without the need for a thermal sintering process. Second, the ALD process can in situ functionalize the Ag NWs to yield defect-free (in terms of blocking gas permeation) coverage of the ALD HZO over the entire nanowire surfaces, which also enhances the ALD-induced fusing of Ag NWs. The composite HZO/Ag NWs films exhibit low sheet resistance (15 Ω sq-1), low water vapor transmission rate (WVTR) (5.1 × 10-6 g m-2 day-1), high optical transmission (92%), excellent flexibility (12.5 mm bending radius), high stability/durability (against an extensive set of degradation modes and photolithographic patterning processes), and low processing temperature (90 °C) and can be used in perovskite solar cells to obtain high power conversion efficiency (14.46%).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-tert-Butylpyridine, 98%