Merck
A porous collagen-carboxymethyl cellulose/hydroxyapatite composite for bone tissue engineering by bi-molecular template method.
International journal of biological macromolecules (2019-06-21)
Xichan He, Keyong Tang, Xiumin Li, Fang Wang, Jie Liu, Fangfang Zou, Mengyuan Yang, Meixuan Li
ABSTRAKT
Inspired by the mechanism of bone formation, a porous collagen-carboxymethyl cellulose/hydroxyapatite (Col-CMC/HA) composite was designed and fabricated using a biomimetic template of Col and CMC protein-polysaccharide bi-molecules. The morphology, composition and physical properties of Col-CMC/HA composites were characterized systematically. It was found that the nano-HA homogenously distributed on the surface of Col-CMC bi-templates while the composite presented 3D porous structure with pore size from 100 μm to 300 μm. The porosities of composites were located at the range of 71%-85%. Besides, the compressive strength of composites was highly depended on the ratio of Col to CMC in the organic template. The optimized composite in respect to physical properties showed a compressive strength as high as 7.06 MPa, quite close to that of natural bone. The high relative growth rate of wild-type mouse embryonic fibroblasts cells was found for the composite, indicating a good biocompatibility. The organic-inorganic composite also behaved good in collagenase resistance and could be biodegraded in 8 weeks, with about 50% of initial weight left at the ratio of Col to CMC of 1:9. The results demonstrated that the Col-CMC/HA composite by bi-molecular template method was a rational and safe method to prepare biomaterials with tunable properties.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)