Merck
  • Derivation and external validation of a model to predict 2-year mortality risk of patients with advanced schistosomiasis after discharge.
Derivation and external validation of a model to predict 2-year mortality risk of patients with advanced schistosomiasis after discharge.
EBioMedicine (2019-08-28)
Guo Li, Shanshan Huang, Lifei Lian, Xiaoyan Song, Wenzhe Sun, Jinfeng Miao, Bohan Li, Yong Yuan, Shengfan Wu, Xiaoyan Liu, Zhou Zhu
ABSTRAKT
To date, no risk prediction tools have been developed to identify high mortality risk of patients with advanced schistosomiasis within 2 years after discharge. We aim to derive and validate a risk prediction model to be applied in clinical practice. The risk prediction model was derived from 1487 patients from Jingzhou and externally validated by 723 patients of Huangshi, two prefecture-level cities in Hubei province, China (from September 2014 to January 2015, with follow-up to January 2017). The baseline variables were collected. The mean age [SD] was 62.89 [10.38] years for the derivation cohort and 62.95 [12.22] years for the external validation cohort. The females accounted for 36.3% and 43.7% of the derivation and validation cohorts, respectively. 8.27% patients (123/1487) in the derivation cohort and 7.75% patients (56/723) in the external validation cohort died within 2 years after discharge. We constructed 4 models based on the 7 selected variables: age, clinical classification, serum direct bilirubin (DBil), aspartate aminotransferase (AST), alkaline phosphatase (ALP), hepatitis B surface antigen (HBsAg), alpha fetoprotein (AFP) at admission. In the external validation cohort, the multivariate model including 7 variables had a C statistic of 0.717 (95% CI, 0.646-0.788) and improved integrated discrimination improvement (IDI) value and net reclassification improvement (NRI) value compared to the other reduced models. Therefore, a multivariate model was developed to predict the 2-year mortality risk for patients with advanced schistosomiasis after discharge. It could also help guide follow-up, aid prognostic assessment and inform resource allocation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)