Merck
  • Detection and quantification of alphaS1-, alphaS2-, beta-, kappa-casein,alpha-lactalbumin, beta-lactoglobulin and lactoferrin in bovine milk by reverse-phase high-performance liquid chromatography
Detection and quantification of alphaS1-, alphaS2-, beta-, kappa-casein,alpha-lactalbumin, beta-lactoglobulin and lactoferrin in bovine milk by reverse-phase high-performance liquid chromatography
Agriculturae Conspectus Scientificus (2013)
Maurmayr A, et al.