Merck
The plasma membrane of bloodstream-form African trypanosomes confers susceptibility and specificity to killing by hydrophobic peptides.
The Journal of biological chemistry (2010-07-10)
John M Harrington, Justin Widener, Natalie Stephens, Thomas Johnson, Maria Francia, Paul Capewell, Annette Macleod, Stephen L Hajduk
ABSTRAKT
Trypanosoma brucei is the causative agent of both a veterinary wasting disease and human African trypanosomiasis, or sleeping sickness. The cell membrane of the developmental stage found within the mammalian host, the bloodstream form (BSF), is highly dynamic, exhibiting rapid rates of endocytosis and lateral flow of glycosylphosphatidylinositol-anchored proteins. Here, we show that the cell membrane of these organisms is a target for killing by small hydrophobic peptides that increase the rigidity of lipid bilayers. Specifically, we have derived trypanocidal peptides that are based upon the hydrophobic N-terminal signal sequences of human apolipoproteins. These peptides selectively partitioned into the plasma membrane of BSF trypanosomes, resulting in an increase in the rigidity of the bilayer, dramatic changes in cell motility, and subsequent cell death. No killing of the developmental stage found within the insect midgut, the procyclic form, was observed. Additionally, the peptides exhibited no toxicity toward mammalian cell lines and did not induce hemolysis. Studies with model liposomes indicated that bilayer fluidity dictates the susceptibility of membranes to manipulation by hydrophobic peptides. We suggest that the composition of the BSF trypanosome cell membrane confers a high degree of fluidity and unique susceptibility to killing by hydrophobic peptides and is therefore a target for the development of trypanocidal drugs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
17:0 PC, 1,2-diheptadecanoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
17:0 PC, 1,2-diheptadecanoyl-sn-glycero-3-phosphocholine, powder
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
15:0 PC, 1,2-dipentadecanoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
18:0-18:1 PC, 1-stearoyl-2-oleoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
18:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder
Avanti
15:0 PC, 1,2-dipentadecanoyl-sn-glycero-3-phosphocholine, powder