Merck
EGFR nuclear translocation modulates DNA repair following cisplatin and ionizing radiation treatment.
Cancer research (2011-01-27)
Gianmaria Liccardi, John A Hartley, Daniel Hochhauser
ABSTRAKT
Epidermal growth factor receptor (EGFR) overexpression is associated with resistance to chemotherapy and radiotherapy. It modulates DNA repair after radiation-induced damage through association with the catalytic subunit of DNA protein kinase (DNA-PKcs). We investigated the role of EGFR nuclear import and its association with DNA-PKcs on DNA repair after exposure to cisplatin or ionizing radiation (IR). The model system was based on EGFR-null murine NIH3T3 fibroblasts in which EGFR expression was restored with isoforms that were wild-type (wt), derived from human cancers (L858R, EGFRvIII), or mutated in the nuclear localization signal (NLS) sequence. In cells expressing wtEGFR or EGFRvIII, there was complete unhooking of cisplatin-induced interstrand cross-links and repair of IR-induced strand breaks. In contrast, cells expressing L858R or NLS mutations showed reduced unhooking of interstrand cross-links and repair of strand breaks. Immunoprecipitation showed wtEGFR and EGFRvIII binding to DNA-PKcs, increasing 2-fold 18 hours after cisplatin therapy. Confocal microscopy and proximity ligation assay showed that this interaction in the cytoplasm and nucleus was associated with increased DNA protein kinase complex (DNA-PK) activity. Cells expressing the EGFR L858R mutation, which has constitutive kinase activity, exhibited reduced DNA repair without nuclear localization. EGFR-NLS mutants showed impaired nuclear localization and DNA-PKcs association with reduced DNA repair and DNA-PK kinase activity. In summary, EGFR nuclear localization was required for modulation of cisplatin and IR-induced repair of DNA damage. EGFR-DNA-PKcs binding was induced by cisplatin or IR but not by EGFR nuclear translocation per se. Our findings show that EGFR subcellular distribution can modulate DNA repair kinetics, with implications for design of EGFR-targeted combinational therapies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.