Merck
Enhanced mGlu5 Signaling in Excitatory Neurons Promotes Rapid Antidepressant Effects via AMPA Receptor Activation.
Neuron (2019-08-20)
Amrei Holz, Felix Mülsch, Martin K Schwarz, Michael Hollmann, Mate D Döbrössy, Volker A Coenen, Marlene Bartos, Claus Normann, Knut Biber, Dietrich van Calker, Tsvetan Serchov
ABSTRAKT
Conventional antidepressants have limited efficacy and many side effects, highlighting the need for fast-acting and specific medications. Induction of the synaptic protein Homer1a mediates the effects of different antidepressant treatments, including the rapid action of ketamine and sleep deprivation (SD). We show here that mimicking Homer1a upregulation via intravenous injection of cell-membrane-permeable TAT-Homer1a elicits rapid antidepressant effects in various tests. Similar to ketamine and SD, in vitro and in vivo application of TAT-Homer1a enhances mGlu5 signaling, resulting in increased mTOR pathway phosphorylation, and upregulates synaptic AMPA receptor expression and activity. The antidepressant action of SD and Homer1a induction depends on mGlu5 activation specifically in excitatory CaMK2a neurons and requires enhanced AMPA receptor activity, translation, and trafficking. Moreover, our data demonstrate a pronounced therapeutic potential of different TAT-fused peptides that directly modulate mGlu5 and AMPA receptor activity and thus might provide a novel strategy for rapid and effective antidepressant treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Homer1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Tetrapropylammonium perruthenate, 97%
Sigma-Aldrich
Anti-GluR1-NT (NT) Antibody, clone RH95, clone RH95, from mouse
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Metabotropic Glutamate Receptor 5 Antibody, pain, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse