Merck
Selective coexpression of multiple chemical markers defines discrete populations of neocortical GABAergic neurons.
Cerebral cortex (New York, N.Y. : 1991) (2011-01-12)
Yoshiyuki Kubota, Naoki Shigematsu, Fuyuki Karube, Akio Sekigawa, Satoko Kato, Noboru Yamaguchi, Yasuharu Hirai, Mieko Morishima, Yasuo Kawaguchi
ABSTRAKT
Whether neocortical γ-aminobutyric acid (GABA) cells are composed of a limited number of distinct classes of neuron, or whether they are continuously differentiated with much higher diversity, remains a contentious issue for the field. Most GABA cells of rat frontal cortex have at least 1 of 6 chemical markers (parvalbumin, calretinin, alpha-actinin-2, somatostatin, vasoactive intestinal polypeptide, and cholecystokinin), with each chemical class comprising several distinct neuronal subtypes having specific physiological and morphological characteristics. To better clarify GABAergic neuron diversity, we assessed the colocalization of these 6 chemical markers with corticotropin-releasing factor (CRF), neuropeptide Y (NPY), the substance P receptor (SPR), and nitric oxide synthase (NOS); these 4 additional chemical markers suggested to be expressed diversely or specifically among cortical GABA cells. We further correlated morphological and physiological characteristics of identified some chemical subclasses of inhibitory neurons. Our results reveal expression specificity of CRF, NPY, SPR, and NOS in morphologically and physiologically distinct interneuron classes. These observations support the existence of a limited number of functionally distinct subtypes of GABA cells in the neocortex.