Merck
Beta(2)-microglobulin identified as an apoptosis-inducing factor and its characterization.
Blood (1999-10-09)
M Mori, Y Terui, M Ikeda, H Tomizuka, M Uwai, T Kasahara, N Kubota, T Itoh, Y Mishima, M Douzono-Tanaka, M Yamada, S Shimamura, J Kikuchi, Y Furukawa, Y Ishizaka, K Ikeda, H Mano, K Ozawa, K Hatake
ABSTRAKT
Major histocompatibility complex (MHC) molecules play an important role in antigen presentation for induction of tumor as well as cellular and humoral immunities. Recent studies using anti-MHC antibodies demonstrated that antibodies specific for HLA class I molecules induced cellular activation and a type of apoptosis that may be distinct from Fas-dependent or TNFR (tumor necrosis factor-alpha receptor)-dependent processes. We purified a previously untested apoptosis-inducing factor from HL-60 human leukemic cell-conditioned media to homogeneity and sequenced it. It was identified as beta(2)-microglobulin (beta(2)m), which has been previously known as thymotaxin and is a part of the HLA class I antigen complex. beta(2)m acts on both T-leukemic cells and myeloid leukemic cells to induce apoptosis, which then activates caspase 1 and 3. Cross-linking studies showed that biotinilated beta(2)m recognized an epitope distinct from those recognized by the anti-HLA class I antibody, as reported previously. We demonstrated that beta(2)m plays a previously unrecognized and important role in regulating the elimination of tumor cells, which occurs as a result of the action of beta(2)m as an apoptosis-inducing factor.