Merck
PIII /PV =O Catalyzed Cascade Synthesis of N-Functionalized Azaheterocycles.
Angewandte Chemie (International ed. in English) (2019-12-24)
Trevor V Nykaza, Gen Li, Junyu Yang, Michael R Luzung, Alexander T Radosevich
ABSTRAKT
An organocatalytic method for the modular synthesis of diverse N-aryl and N-alkyl azaheterocycles (indoles, oxindoles, benzimidazoles, and quinoxalinediones) is reported. The method employs a small-ring organophosphorus-based catalyst (1,2,2,3,4,4-hexamethylphosphetane P-oxide) and a hydrosilane reductant to drive the conversion of ortho-functionalized nitroarenes into azaheterocycles through sequential intermolecular reductive C-N cross coupling with boronic acids, followed by intramolecular cyclization. This method enables the rapid construction of azaheterocycles from readily available building blocks, including a regiospecific approach to N-substituted benzimidazoles and quinoxalinediones.