Merck
Uncovering Cellular retinoic acid-binding protein 2 as a potential target for rheumatoid arthritis synovial hyperplasia.
Scientific reports (2018-06-09)
Nerea Mosquera, Angela Rodriguez-Trillo, Antonio Mera-Varela, Antonio Gonzalez, Carmen Conde
ABSTRAKT
Rheumatoid arthritis (RA) is a systemic autoimmune disease including synovitis and synovial hyperplasia that contribute to joint destruction. Pivotal pathogenic mechanisms in this process are the dysregulated proliferation and apoptosis of fibroblast-like synoviocytes (FLS). Unfortunately, the mechanisms of FLS dysregulation are not completely elucidated. Here, we explored a new hypothesis based in the potent anti-proliferative and pro-apoptotic activity of retinoids in some types of cancer. Specifically, we investigated the role of retinoids and of the retinoic acid binding proteins, CRABP2 and FABP5, on the proliferation and apoptosis of FLS from RA by adding all-trans retinoic acid (ATRA) or silencing CRABP2 and FABP5. We showed an unconventional behaviour of RA FLS, which were relatively insensitive to ATRA. In effect, ATRA increased the resistance to apoptosis despite the high CRABP2/FABP5 ratio of RA FLS; and CRABP2 suppression sensitized RA FLS to Fas-induced apoptosis. This latter effect was associated with changes in expression of kinases, ASK1 up-regulation and ERK down-regulation, and increased phosphorylation of JNK. In addition, the potentiation of FLS apoptosis by CRABP2 silencing persisted in the presence of pro-inflammatory mediators, TNF e IL1β. Therefore, the results point to CRABP2 as a potential target to decrease synovial hyperplasia in RA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid