Merck
The lincRNA MIRAT binds to IQGAP1 and modulates the MAPK pathway in NRAS mutant melanoma.
Scientific reports (2018-07-22)
Martina Sanlorenzo, Igor Vujic, Rosaura Esteve-Puig, Kevin Lai, Marin Vujic, Kevin Lin, Christian Posch, Michelle Dimon, Adrian Moy, Mitchell Zekhtser, Katia Johnston, Deborah Gho, Wilson Ho, Abhinay Gajjala, Juan Oses Prieto, Alma Burlingame, Adil Daud, Klemens Rappersberger, Susana Ortiz-Urda
ABSTRAKT
Despite major advances in targeted melanoma therapies, drug resistance limits their efficacy. Long noncoding RNAs (lncRNAs) are transcriptome elements that do not encode proteins but are important regulatory molecules. LncRNAs have been implicated in cancer development and response to different therapeutics and are thus potential treatment targets; however, the majority of their functions and molecular interactions remain unexplored. In this study, we identify a novel cytoplasmic intergenic lincRNA (MIRAT), which is upregulated following prolonged MAPK inhibition in NRAS mutant melanoma and modulates MAPK signaling by binding to the MEK scaffold protein IQGAP1. Collectively, our results present MIRAT's direct modulatory effect on the MAPK pathway and highlight the relevance of cytoplasmic lncRNAs as potential targets in drug resistant cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-IQGAP1, klon AF4, clone AF4, Upstate®, from mouse