Merck
Maternal diabetes promotes mTORC1 downstream signalling in rabbit preimplantation embryos.
Reproduction (Cambridge, England) (2016-02-03)
Jacqueline Gürke, Maria Schindler, S Mareike Pendzialek, René Thieme, Katarzyna J Grybel, Regine Heller, Katrin Spengler, Tom P Fleming, Bernd Fischer, Anne Navarrete Santos
ABSTRAKT
The mammalian target of rapamycin complex 1 (mTORC1) is known to be a central cellular nutrient sensor and master regulator of protein metabolism; therefore, it is indispensable for normal embryonic development. We showed previously in a diabetic pregnancy that embryonic mTORC1 phosphorylation is increased in case of maternal hyperglycaemia and hypoinsulinaemia. Further, the preimplantation embryo is exposed to increased L-leucine levels during a diabetic pregnancy. To understand how mTOR signalling is regulated in preimplantation embryos, we examined consequences of L-leucine and glucose stimulation on mTORC1 signalling and downstream targets in in vitro cultured preimplantation rabbit blastocysts and in vivo. High levels of L-leucine and glucose lead to higher phosphorylation of mTORC1 and its downstream target ribosomal S6 kinase 1 (S6K1) in these embryos. Further, L-leucine supplementation resulted in higher embryonic expression of genes involved in cell cycle (cyclin D1; CCND1), translation initiation (eukaryotic translation initiation factor 4E; EIF4E), amino acid transport (large neutral amino acid transporter 2; Lat2: gene SLC7A8) and proliferation (proliferating cell nuclear antigen; PCNA) in a mTORC1-dependent manner. Phosphorylation of S6K1 and expression patterns of CCND1 and EIF4E were increased in embryos from diabetic rabbits, while the expression of proliferation marker PCNA was decreased. In these embryos, protein synthesis was increased and autophagic activity was decreased. We conclude that mammalian preimplantation embryos sense changes in nutrient supply via mTORC1 signalling. Therefore, mTORC1 may be a decisive mediator of metabolic programming in a diabetic pregnancy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder