Merck
  • Endotoxin attenuates growth hormone-induced hepatic insulin-like growth factor I expression by inhibiting JAK2/STAT5 signal transduction and STAT5b DNA binding.
Endotoxin attenuates growth hormone-induced hepatic insulin-like growth factor I expression by inhibiting JAK2/STAT5 signal transduction and STAT5b DNA binding.
American journal of physiology. Endocrinology and metabolism (2007-03-01)
Yu Chen, Difei Sun, Vidya M R Krishnamurthy, Ralph Rabkin
ABSTRAKT
Gram-negative sepsis with release of endotoxin is a frequent cause of cachexia that develops partly because of resistance to growth hormone (GH) with reduced insulin-like growth factor-I (IGF-I) expression. We set out to more fully characterize the mechanisms for the resistance and to determine whether in addition to a defect in the janus kinase 2 (JAK2)-signal transducer and activator of transcription (STAT) 5b pathway, required for GH-induced IGF-I expression, there might also be a more distal defect. Conscious rats were given endotoxin and studied 4 h later. In liver of these animals, GH-induced JAK2 and STAT5 phosphorylation was impaired and appeared to be caused, at least in part, by a marked increase in hepatic tumor necrosis factor-alpha and interleukin-6 mRNA expression accompanied by elevated levels of inhibitors of GH signaling, namely cytokine-inducible suppressors of cytokine signaling-1 and -3 and cytokine-inducible SH2 protein (CIS). Nuclear phosphorylated STAT5b levels were significantly depressed to 61% of the control values and represent a potential cause of the reduced GH-induced IGF-I expression. In addition, binding of phosphorylated STAT5b to DNA was reduced to an even greater extent and averaged 17% of the normal control value. This provides a further explanation for the impaired IGF-I gene transcription. Interestingly, when endotoxin-treated rats were treated with GH, there was a marked increase in proinflammatory cytokine gene expression in the liver. If such a response were to occur in humans, this might provide a partial explanation for the adverse effect of GH treatment reported in critically ill patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-fosfo-STAT5A/B (Tyr694/699), klon 8-5-2, clone 8-5-2, Upstate®, from mouse