Merck
The ABD on the nascent polypeptide and PH domain are required for the precise Anillin localization in Drosophila syncytial blastoderm.
Scientific reports (2018-08-29)
Tomoki Hirashima, Ryo Tanaka, Masamitsu Yamaguchi, Hideki Yoshida
ABSTRAKT
Targeting proteins to regions where they are required is essential for proper development of organisms. For achievement of this, subcellular mRNA localization is one of the critical mechanisms. Subcellular mRNA localization is an evolutionarily conserved phenomenon from E. coli to human and contributes to limiting the regions at which its products function and efficiently supplies substrates for protein translation. During early Drosophila embryogenesis, while 71% of the 3370 mRNAs analyzed have shown prominent subcellular localization, the underlying molecular mechanisms have not been elucidated. Here, we reveal that anillin mRNA, one of the localized mRNAs in early Drosophila embryo, localizes to the tip of the pseudo-cleavage furrow in the Drosophila syncytial blastoderm using in situ hybridization combined with immunohistochemistry. Localization analyses with transgenic fly lines carrying a series of deletion mRNAs indicate that this localization is dependent on its own nascent polypeptides including the actin binding domain (ABD). In addition to the mRNA localization, it is revealed that the pleckstrin homology (PH) domain of Anillin protein is also required for its proper localization. Thus, we indicate that the precise localization of Anillin protein is tightly regulated by the ABD on the nascent polypeptide and PH domain in the Drosophila syncytial blastoderm.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2