Merck
  • Ceramide generated by acidic sphingomyelinase contributes to tumor necrosis factor-alpha-mediated apoptosis in human colon HT-29 cells through glycosphingolipids formation. Possible role of ganglioside GD3.
Ceramide generated by acidic sphingomyelinase contributes to tumor necrosis factor-alpha-mediated apoptosis in human colon HT-29 cells through glycosphingolipids formation. Possible role of ganglioside GD3.
FEBS letters (2002-09-05)
Anna Colell, Albert Morales, José C Fernández-Checa, Carmen García-Ruiz
ABSTRAKT
In the present study we assessed the contribution of acidic sphingomyelinase (ASMase), a ceramide generating enzyme, in tumor necrosis factor (TNF)-mediated apoptosis in human colon HT-29 cells. TNF induced apoptosis in HT-29 cells in a time- and dose-dependent fashion. Downregulation of the active endogenous ASMase form prevented TNF-stimulated ASMase activity and apoptosis. Furthermore, inhibition of glucosylceramide synthase, which blunted TNF-stimulated GD3 levels, abolished TNF-mediated cell death. Immunocytochemical staining revealed the co-localization of GD3 with mitochondria induced by TNF. The knockdown of targeted GD3 synthase by antisense expression vector protected HT-29 cells against TNF-induced cell death. Thus, ASMase plays a key role in TNF-induced cell death in human colon epithelial cells possibly through GD3 generation.