Merck
A novel YAP1/SLC35B4 regulatory axis contributes to proliferation and progression of gastric carcinoma.
Cell death & disease (2019-06-09)
Jun Liu, Xinhui Zhao, Ke Wang, Xiang Zhang, Yanping Yu, Yongzhi Lv, Shun Zhang, Long Zhang, Yuan Guo, Yunlong Li, Angang Yang, Rui Zhang, Jipeng Li
ABSTRAKT
Solute carrier family 35 member B4 (SLC35B4), a nucleotide sugar transporter, is capable of transporting UDP-xylose and UDP-GlcNAc from the cytoplasm to the lumen of the endoplasmic reticulum and Golgi. SLC35B4 has a pivotal role in glycosylation of biological macromolecules. However, its functional roles and regulatory mechanisms in malignant diseases remain unknown. Here, using the cDNA arrays, promoter reporter assays, and chromatin immunoprecipitation assays, we demonstrated that SLC35B4 is directly transactivated by YAP1-TEADs complex in gastric cancer (GC) cells. CCK-8, plate colony formation and soft agar assays revealed that SLC35B4 is essential for survival and proliferation in GC cells and nude mice models. SLC35B4 expression is markedly higher in GC tissues compared with control noncancerous tissues. Immunohistochemistry revealed that SLC35B4 expression is positively correlated with YAP1 expression in human GC tissues, and this correlation is also confirmed in the GC TCGA data set. GC patients with high levels of SLC35B4 expression have poorer prognosis than those with low levels of SLC35B4 expression. Collectively, our findings defined SLC35B4 as an important downstream oncogenic target of YAP1, suggesting that dysregulated signaling of a novel YAP1/SLC35B4 axis promotes GC development and progression, and this axis could be a potential candidate for prognosis and therapeutics in GC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-SLC35B4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid