Merck
  • Characterization of nine microsatellite loci on endemic kangaroo rats Dipodomys simulans peninsularis from southern Baja California Peninsula.
Characterization of nine microsatellite loci on endemic kangaroo rats <I>Dipodomys simulans peninsularis</I> from southern Baja California Peninsula.
Molecular Ecology Notes (2007)
Vega, A.M., et al.