Merck
Panels of cytokines and other secretory proteins as potential biomarkers of ovarian endometriosis.
The Journal of molecular diagnostics : JMD (2015-03-24)
Vida Kocbek, Katja Vouk, Nick A Bersinger, Michael D Mueller, Tea Lanišnik Rižner
ABSTRAKT
Endometriosis is a gynecologic disease that is characterized by nonspecific symptoms and invasive diagnostics. To date, there is no adequate noninvasive method for the diagnosis of endometriosis. Although more than 100 potential biomarkers have been investigated in blood and/or peritoneal fluid, none of these has proven useful in clinical practice. The aim to find a suitable panel of biomarkers that would allow noninvasive diagnosis thus remains of interest. We evaluated the concentrations of 16 cytokines and other secretory proteins in serum and peritoneal fluid of 58 women with ovarian endometriosis (cases) and 40 healthy women undergoing sterilization or patients with benign ovarian cysts (controls) using multiplexed double fluorescence-based immunometric assay platform and enzyme-linked immunosorbent assay. Significantly higher concentrations of glycodelin-A were shown in serum, and significantly higher levels of glycodelin-A, IL-6, and IL-8, and lower levels of leptin were measured in the peritoneal fluid of cases versus controls. In serum, the best performance was shown by models that included the ratio of leptin/glycodelin-A and the ratio of ficolin 2/glycodelin-A, whereas in the peritoneal fluid the best models included the ratio of biglycan/leptin, regulated on activation normal T-cell expressed and secreted/IL-6 and ficolin-2/glycodelin-A, and IL-8 per milligram of total protein, all in combination with age. The models using serum and peritoneal fluid distinguished between ovarian endometriosis patients and controls regardless of the menstrual cycle phase with relatively high sensitivity (72.5% to 84.2%), specificity (78.4% to 91.2%), and area under the curve (0.85 to 0.90).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-BGN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-C-Reactive Protein antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder