Merck
Non-canonical translation initiation in yeast generates a cryptic pool of mitochondrial proteins.
Nucleic acids research (2019-06-20)
Geoffray Monteuuis, Anna Miścicka, Michał Świrski, Lounis Zenad, Olli Niemitalo, Lidia Wrobel, Jahangir Alam, Agnieszka Chacinska, Alexander J Kastaniotis, Joanna Kufel
ABSTRAKT
Utilization of non-AUG alternative translation start sites is most common in bacteria and viruses, but it has been also reported in other organisms. This phenomenon increases proteome complexity by allowing expression of multiple protein isoforms from a single gene. In Saccharomyces cerevisiae, a few described cases concern proteins that are translated from upstream near-cognate start codons as N-terminally extended variants that localize to mitochondria. Using bioinformatics tools, we provide compelling evidence that in yeast the potential for producing alternative protein isoforms by non-AUG translation initiation is much more prevalent than previously anticipated and may apply to as many as a few thousand proteins. Several hundreds of candidates are predicted to gain a mitochondrial targeting signal (MTS), generating an unrecognized pool of mitochondrial proteins. We confirmed mitochondrial localization of a subset of proteins previously not identified as mitochondrial, whose standard forms do not carry an MTS. Our data highlight the potential of non-canonical translation initiation in expanding the capacity of the mitochondrial proteome and possibly also other cellular features.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution