Merck
  • Glycovariant anti-CD37 monospecific protein therapeutic exhibits enhanced effector cell-mediated cytotoxicity against chronic and acute B cell malignancies.
Glycovariant anti-CD37 monospecific protein therapeutic exhibits enhanced effector cell-mediated cytotoxicity against chronic and acute B cell malignancies.
mAbs (2013-07-26)
Sarwish Rafiq, Anthony Siadak, Jonathan P Butchar, Carolyn Cheney, Gerard Lozanski, Naduparambil K Jacob, Rosa Lapalombella, Jackie McGourty, Meghan Moledor, Richard Lowe, Ben Setter, Jeffrey Jones, Joseph M Flynn, Leslie Andritsos, Steven Devine, Xiaokui Mo, David Jarjoura, Susheela Tridandapani, Paul Algate, John C Byrd, Natarajan Muthusamy
ABSTRAKT
TRU-016 is a SMIP(TM) (monospecific protein therapeutic) molecule against the tetraspanin transmembrane family protein CD37 that is currently in Phase 2 trials in Chronic Lymphocytic Leukemia (CLL) and Non-Hodgkin Lymphoma (NHL). In an attempt to enhance the ADCC function of SMIP-016, the chimeric version of TRU-016, SMIP-016(GV) was engineered with a modification in a glycosylation site in the Fc domain. The wild-type and glycovariant SMIP proteins mediate comparable Type I antibody-like direct cytotoxicity in the presence of anti-human Fc crosslinker and show a similar tyrosine phosphorylation pattern post-treatment. However, NK cells stimulated with the SMIP-016(GV) exhibit enhanced activation and release 3-fold more interferon-γ compared with SMIP-016. SMIP-016(GV) shows enhanced ADCC function against cells expressing CD37 with NK cell effectors derived from both normal and CLL-affected individuals. Enhanced ADCC is observed against CLL cells and is sustained at concentrations of SMIP-016(GV) as low at 5E(-6) µg/mL on cells expressing minimal CD37 antigen. In support of the biological relevance of this, SMIP-016(GV) mediates effective ADCC against primary acute lymphoblastic leukemia (ALL) cells with low surface expression of CD37. Collectively, these data suggest potential use of the novel therapeutic agent SMIP-016(GV) with enhanced effector function for B cell malignancies, including CLL and ALL therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
PKH67 Green Fluorescent Cell Linker Midi Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Przeciwciało przeciw fosfotyrozynie, klon ®, clone 4G10®, Upstate®, from mouse