Merck
A High-Resolution Map of SBP1 Interactomes in Plasmodium falciparum-infected Erythrocytes.
iScience (2019-09-03)
Ryo Takano, Hiroko Kozuka-Hata, Daisuke Kondoh, Hiroki Bochimoto, Masaaki Oyama, Kentaro Kato
ABSTRAKT
The pathogenesis of malaria parasites depends on host erythrocyte modifications that are facilitated by parasite proteins exported to the host cytoplasm. These exported proteins form a trafficking complex in the host cytoplasm that transports virulence determinants to the erythrocyte surface; this complex is thus essential for malaria virulence. Here, we report a comprehensive interaction network map of this complex. We developed authentic, unbiased, highly sensitive proteomic approaches to determine the proteins that interact with a core component of the complex, SBP1 (skeleton-binding protein 1). SBP1 interactomes revealed numerous exported proteins and potential interactors associated with SBP1 intracellular trafficking. We identified several host-parasite protein interactions and linked the exported protein MAL8P1.4 to Plasmodium falciparum virulence in infected erythrocytes. Our study highlights the complicated interplay between parasite and host proteins in the host cytoplasm and provides an interaction dataset connecting dozens of exported proteins required for P. falciparum virulence.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-STOM antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Spectrin (α and β) antibody produced in mouse, clone SB-SP1, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)