Merck
The protective variant rs7173049 at LOXL1 locus impacts on retinoic acid signaling pathway in pseudoexfoliation syndrome.
Human molecular genetics (2019-04-16)
Daniel Berner, Ursula Hoja, Matthias Zenkel, James Julian Ross, Steffen Uebe, Daniela Paoli, Paolo Frezzotti, Robyn M Rautenbach, Ari Ziskind, Susan E Williams, Trevor R Carmichael, Michele Ramsay, Fotis Topouzis, Anthi Chatzikyriakidou, Alexandros Lambropoulos, Periasamy Sundaresan, Humaira Ayub, Farah Akhtar, Raheel Qamar, Juan C Zenteno, Marisa Cruz-Aguilar, Yury S Astakhov, Michael Dubina, Janey Wiggs, Mineo Ozaki, Friedrich E Kruse, Tin Aung, André Reis, Chiea Chuen Khor, Francesca Pasutto, Ursula Schlötzer-Schrehardt
ABSTRAKT
LOXL1 (lysyl oxidase-like 1) has been identified as the major effect locus in pseudoexfoliation (PEX) syndrome, a fibrotic disorder of the extracellular matrix and frequent cause of chronic open-angle glaucoma. However, all known PEX-associated common variants show allele effect reversal in populations of different ancestry, casting doubt on their biological significance. Based on extensive LOXL1 deep sequencing, we report here the identification of a common noncoding sequence variant, rs7173049A>G, located downstream of LOXL1, consistently associated with a decrease in PEX risk (OR=0.63, p=6.33x10-31) in nine different ethnic populations. We provide experimental evidence for a functional enhancer-like regulatory activity of the genomic region surrounding rs7173049 influencing expression levels of ISLR2 (immunoglobulin superfamily containing leucine-rich repeat protein 2) and STRA6 (stimulated by retinoic acid receptor 6), apparently mediated by allele-specific binding of the transcription factor THRβ (thyroid hormone receptor beta). We further show that the protective rs7173049-G allele correlates with increased tissue expression levels of ISLR2 and STRA6 and that both genes are significantly downregulated in tissues of PEX patients together with other key components of the STRA6 receptor-driven retinoic acid signaling pathway. siRNA-mediated downregulation of retinoic acid signaling induces upregulation of LOXL1 and PEX-associated matrix genes in PEX-relevant cell types. These data indicate that dysregulation of STRA6 and impaired retinoid metabolismare involved in the pathophysiology of PEX syndrome and that the variant rs7173049-G, which represents the first common variant at the broad LOXL1 locus without allele effect reversal, mediates a protective effect through upregulation of STRA6 in ocular tissues.