Merck
  • Conjugation reactions in the preparations of quantum dot-based immunoluminescent probes for analysis of proteins by capillary electrophoresis.
Conjugation reactions in the preparations of quantum dot-based immunoluminescent probes for analysis of proteins by capillary electrophoresis.
Analytical and bioanalytical chemistry (2011-02-08)
M Lišková, I Voráčová, K Klepárník, V Hezinová, J Přikryl, F Foret
ABSTRAKT
A number of biologically important molecules, such as DNA, proteins, and antibodies, are routinely conjugated with fluorescent tags for high-sensitivity analyses. Here, the application of quantum dots in the place of bright and size-tunable luminophores is studied. Several selected bioconjugation reactions via zero-length cross-linkers, long-chain linkers, and oriented methods for linking of quantum dots with proteins were tested. Anti-ovalbumin, anti-proliferating cell nuclear antigen, anti-hemagglutinin, and anti-CD3 membrane protein as model antibodies and annexin V were used as high-specificity selectors. The reaction yield and efficiency of the prepared immunoluminescent probes were tested by capillary zone electrophoresis with laser-induced fluorescence detection.