Merck
Enhanced expression of IL-34 in an inflammatory cyst of the submandibular gland: a case report.
Inflammation and regeneration (2018-07-14)
Muhammad Baghdadi, Kozo Ishikawa, Hiraku Endo, Yui Umeyama, Tsukasa Ataka, Haruka Wada, Yumiko Oyamada, Naoki Hyakushima, Ken-Ichiro Seino
ABSTRAKT
Cysts of the salivary glands are common lesions that occur in the context of various etiologies. Although the diagnostic importance of cysts in salivary gland diseases has been well studied, molecular mechanisms that control the related pathological process remain largely unknown. IL-34 is a novel cytokine that was discovered recently as a tissue-specific ligand of colony stimulating factor-1 receptor. Since its discovery, accumulating evidence has revealed emerging roles of IL-34 in various pathological conditions and has been suggested to correlate remarkably with inflammation. In this study, we report a medical case of an inflammatory cyst within the submandibular gland, through which evaluating the possible involvement of IL-34 in salivary gland disorders. A 37-year-old male patient suffered from a sudden swelling in the right submandibular region, started initially small and had gradually increased in size to reach 3-4 cm in 1 week, accompanied by pain and local fever. Ultrasonography and MRI imaging revealed the existence of a well-defined cystic lesion with sharp borders measuring 39.8 mm × 19.7 mm within the right submandibular gland. The cyst was removed surgically, and the diagnostic decision was determined based on histopathological observations as an inflammatory cyst in the submandibular gland. Sections were generated from different regions of the surgically resected inflammatory cyst and used to examine IL-34 expression by immunohistochemistry compared to normal salivary gland tissues. Immunohistochemical staining showed enhanced expression of IL-34 in the ductal epithelial cells and endothelial cells of blood vessels, with a tendency to be accompanied with high infiltration of immune cells, which suggests a possible involvement of IL-34 in the pathogenesis of salivary gland inflammation. In this report, we introduce interesting findings of enhanced IL-34 expression in a case of an inflamed submandibular gland. Our findings emphasize the pathological roles of IL-34 as an inflammation amplifier and angiogenic enhancer in inflammatory conditions, such as in salivary gland disorders.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-IL-34, klon 1D12, ascites fluid, clone 1D12, from mouse