Merck
Expression of long-term depression underlies visual recognition memory.
Neuron (2008-04-29)
Sarah Griffiths, Helen Scott, Colin Glover, Alison Bienemann, Mohamed T Ghorbel, James Uney, Malcolm W Brown, E Clea Warburton, Zafar I Bashir
ABSTRAKT
The modifications occurring in the brain during learning and memory are still poorly understood but may involve long-lasting changes in synaptic transmission (synaptic plasticity). In perirhinal cortex, a lasting decrement in neuronal responsiveness is associated with visual familiarity discrimination, leading to the hypothesis that long-term depression (LTD)-like synaptic plasticity may underlie recognition memory. LTD relies on internalization of AMPA receptors (AMPARs) through interaction between their GluR2 subunits and AP2, the clathrin adaptor protein required for endocytosis. We demonstrate that a peptide that blocks interactions between GluR2 and AP2 blocks LTD in perirhinal cortex in vitro. Viral transduction of this peptide in perirhinal cortex produced striking deficits in visual recognition memory. Furthermore, there was a deficit of LTD in perirhinal cortex slices from virally transduced, recognition memory-deficient animals. These results suggest that internalization of AMPA receptors, a process critical for the expression of LTD in perirhinal cortex, underlies visual recognition memory.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse