Merck
Abstracts from USCAP 2019: Neuropathology and Ophthalmic Pathology (1621-1666).
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2019-03-20)
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Histone H3.3 Antibody, K27M mutant, from rabbit, purified by affinity chromatography