Merck
Targeted analysis reveals alteration in pathway in 5p minus individuals.
Biomedical chromatography : BMC (2019-08-07)
Danielle Zildeana Sousa Furtado, Fernando Brunale Vilela de Moura Leite, Leticia Dias Lima Jedlicka, Danilo Santos Souza, Cleber Nunes Barreto, Heron Dominguez Torres da Silva, Nilson Antonio Assunção
ABSTRAKT
Cri du Chat or 5p minus (5p-) syndrome is characterized by a deletion located on the chromosome 5 short (-p) arm and has an incidence rate of 1 in 50,000 individuals worldwide. This disease manifests in disturbances across a range of systems biochemicals. Therefore, a targeted metabolomics analysis was evaluated in patients with 5p- syndrome to help unravel the biochemical changes that occur in this disease. Urine samples were collected from people of both sexes aged 1-38 years old and analyzed by ultra-performance liquid chromatography coupled to mass spectrometry. Student' statistical test, metabolomic pathway analysis and metabolite set enrichment analysis were applied to the data. Alterations of some amino acids and amine biogenics levels were found in Cri du Chat Syndrome individuals. The alteration of most of these metabolites is associated with energy recuperation and glycolysis. In general, we found the catabolism of some metabolic pathways to be affected in 5p- patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Kynurenine, ≥95.0% (NT)
Sigma-Aldrich
DL-2-Aminoadipic acid, ≥99%