Merck
Toxicities of 4,5-Dihydroisoxazoles Against Root-Knot Nematodes and in Silico Studies of Their Modes of Action.
Journal of agricultural and food chemistry (2020-01-08)
Rodrigo M Fráguas, Viviane A Costa, Willian C Terra, Alcino P Aguiar, Samuel J Martins, Vicente P Campos, Denilson F Oliveira
ABSTRAKT
The present work sought to contribute to the development of new nematicides. Benzaldehydes were initially converted to nitrile oxides that underwent 1,3-dipolar cycloaddition reactions with methyl acrylate to generate 4,5-dihydroisoxazoles. In in vitro tests, methyl 3-phenyl-4,5-dihydroisoxazole-5-carboxylate (1) and methyl 3-(4-chlorophenyl)-4,5-dihydroisoxazole-5-carboxylate (4) increased the mortality of Meloidogyne exigua and Meloidogyne incognita second-stage juveniles (J2). Compounds 1 and 4 presented necessary concentrations of 398 and 501 μg mL-1, respectively, to kill 50% of M. incognita J2 (LC50 values), while the value for carbofuran (positive control) was 168 μg mL-1. In in vivo tests, compounds 1 and 4 reduced the number of M. incognita galls in tomato roots by 70 and 40%, respectively, and the number of eggs by 89 and 44%. Using an in silico approach, we showed that compounds 1 and 4 were toxic to the nematodes by binding to the allosteric binding sites of the agonist-binding domains of the nematode nicotinic acetylcholine receptors. These results opened up possibilities for further investigations aimed at developing novel commercial nematicides.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbofuran, 98%