Merck
Preparation of Calcipotriol Emulsion Using Bacterial Exopolysaccharides as Emulsifier for Percutaneous Treatment of Psoriasis Vulgaris.
International journal of molecular sciences (2019-12-22)
Bo Song, Ruiteng Song, Min Cheng, Hairong Chu, Fang Yan, Yuzhen Wang
ABSTRAKT
An exopolysaccharides/calcipotriol (EPS/CPT) emulsion was prepared using bacterial EPS as emulsifier, sunflower oil as an oil phase and CPT as the loaded drug, and the effect of this emulsion on psoriasis vulgaris treatment was evaluated. An EPS composed of mannose (70.56%) and glucose (29.44%) was obtained from the marine mangrove bacteria Bacillus amyloliquefaciens ZWJ (Zhu Wenjing) strain. The EPS has significant emulsifying activity at the concentration of 1.5%. The prepared EPS/CPT emulsion has small and stable particle size, with a drug content of 0.00492%, and good spreading properties. The in vitro drug release results revealed that the emulsion showed a certain sustained release effect. In vitro and in vivo animal experiments show that the EPS/CPT emulsion can effectively treat psoriasis vulgaris by increasing the accumulation of CPT in psoriatic skin lesions and reducing the levels of inflammatory cells and inflammatory factors (TNF and IL6). Additionally, it has a certain effect on reducing the side effects associated with CPT. This study lays a foundation for the research of EPS in the topical application of medical materials and treatment of psoriasis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder