Merck
Expression, purification, and breast cancer cell inhibiting effect of recombinant human lactoferrin C-lobe.
Bioscience, biotechnology, and biochemistry (2015-09-26)
Lulu Hu, Chen-Hui Gao, Chao Hong, Qiao Zhong, Hong-Liang Dong, Xiao-Ming Gao
ABSTRAKT
Lactoferrin (LTF), a multifunctional glycoprotein of the transferrin family mainly found in exotic secretions in mammals, is an important defense molecule against not only microbial invasion but also tumors. It folds into two globular domains (N- and C-lobes) each containing an iron-binding site. The cationic antimicrobial peptide in N-lobe is known to exert anti-tumor effect via a non-receptor-mediated pathway. However, whether LTF C-lobe also contributes to its anti-tumor activity remains to be investigated. In this study, a human LTF fragment (amino acid residues 343-682) covering the C-lobe was expressed with a histidine tag in E. coli and the purified polypeptide refolded through a series of buffer changing procedure. The resultant recombinant protein caused significant growth arrest of breast carcinoma cells MDA-MB-231 in a dose- and time-dependent manner, evidently via induction of apoptosis of the cell. Our data suggest a positive role for the C-lobe of human LTF in controlling tumors in vitro.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Lactoferrin antibody produced in rabbit, fractionated antiserum, lyophilized powder