Merck
Contribution of small GTPase Rho and its target protein rock in a murine model of lung fibrosis.
American journal of respiratory and critical care medicine (2001-02-24)
Y Shimizu, K Dobashi, K Iizuka, T Horie, K Suzuki, H Tukagoshi, T Nakazawa, Y Nakazato, M Mori
ABSTRAKT
Excess fibroblasts and inflammatory cells may play an important role in the pathogenesis of idiopathic pulmonary fibrosis (IPF). The small GTPase, Rho, and its target protein, Rho-associated coiled-coil-forming protein kinase (ROCK), have been recognized to be major regulators of cell locomotion mediated by reorganization of the actin cytoskelton. Activated ROCK inhibits myosin phosphatase, and this in turn induces phosphorylation of the myosin light chain (MLC). To determine the mechanisms underlying the deterioration process of IPF, we investigated the effect of Y-27632, a selective ROCK inhibitor, in a murine model of bleomycin (BLM)-induced lung fibrosis. The Aschcroft score and hydroxyproline content of the BLM-treated mouse lung decreased in response to Y-27632 treatment. The number of broncoalveolar cells was decreased by Y-27632, and migration of macrophages, neutrophils, and fibroblasts in vitro was inhibited by Y-27632 regardless of various stimuli. Although expression of ROCK-II mRNA in the lung homogenates of the BLM-treated mice was increased approximately 9-fold, expression of ROCK-II protein showed only a slight tendency to increase. BLM elevated MLC phosphorylation levels, and Y-27632 inhibited BLM response. These findings indicate that the Rho/ROCK-mediated pathway plays an important role in IPF, and that blocking of this pathway leads to inhibition of IPF development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Myosin (Light Chains 20 kDa) antibody produced in mouse, clone MY-21, ascites fluid