Merck
The Diagnosis and Treatment of Acute Traumatic Bleeding and Coagulopathy.
Deutsches Arzteblatt international (2019-12-19)
Marc Maegele
ABSTRAKT
Uncontrolled bleeding with trauma-induced coagulopathy (TIC) is still the most common avoidable cause of death in multiple trauma. The aging of the population has led to an increasing number of bleeding trauma patients with pre-existing anticoagulation. Such patients are not treated uniformly, even in major trauma centers. This review is based on a selective search of the literature (Medline/PubMed, Cochrane Reviews) and summarizes current treatment recommendations, including those of the newly revised European trauma guidelines. The treatment of traumatic hemorrhage begins at the site of the accident, with compression, tourniquets, pelvic binders, and rapid transport to a certified trauma center. The early use of tourniquets was shown to lessen the trans- fusion requirement (packed red blood cells: 2.0 ± 0.1 vs. 9.3 ± 0.6; p < 0.001; fresh frozen plasma concentrates: 1.4 ± 0.08 vs. 6.2 ± 0.4; p < 0.001), while external pelvic stabilization was shown to reduce mortality (19.1% vs. 33.3%). Upon the patient's arrival in the hospital, steps are taken to measure, monitor, and support clotting function. Bleeding is controlled surgically according to the principles of damage control. Modern clotting management consists of goal-oriented, individualized therapy, including the use of point-of-care viscoelastic test procedures. Idarucizumab can be used as an antidote to the thrombin inhibitor dabigatran, andexanet alpha as an antidote to factor Xa inhibitors. The evidence-based treatment of patients with hemorrhage from severe trauma, in accordance with the existing guidelines, can improve the clinical outcome. Corresponding algorithms, adapted to local logistics and infrastructure, must be developed and implemented.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyridinium chlorochromate, 98%