Merck
Unexpected Synthesis, Properties, and Nonvolatile Memory Device Application of Imidazole-Fused Azaacenes.
The Journal of organic chemistry (2019-10-28)
Kexiang Zhao, Fei Yu, Wenbo Liu, Yinjuan Huang, Ahmed Ali Said, Yang Li, Qichun Zhang
ABSTRAKT
The unexpected synthesis and characterization of imidazole-fused azaacenes are presented. Their optical and electrochemical properties have been investigated and compared with these of previously reported imidazole-fused azaacenes. Application of these two imidazole-fused azaacenes in memory devices showed distinctly different resistive behaviors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propylene sulfite, 99%